Anti-TEKT2 Rabbit Polyclonal Antibody
400 µL
Catalog # 89515-010
Supplier: Abgent
Supplier #: AP19286A
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Western Blot: 1:1000
Type: Primary
Antigen: TEKT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Documents
Specifications
- Size:400 µL
- Clonality:Polyclonal
- Molecular Weight:49.672 kDa
- Purification:Protein A & Peptide affinity purification
- Epitope:N-Terminal
- Storage Buffer:PBS with 0.09% (W/V) sodium azide, pH7.4
- Host:Rabbit
- Sequence:TLRESRRDIDVVKDPC
- Concentration:0.5mg/ml
- Cat. No.:89515-010
- Reactivity:Human
- Western Blot:Yes
- Shipping Temperature:4 °C
- Antibody Type:Primary
- Antigen Symbol:TEKT2
- Gene ID:27285
- Immunogen:This TEKT2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 103-132 amino acids from the N-terminal region of human TEKT2.
- Amino Acid Number:103 to 132
- Conjugation:Unconjugated
- Antigen Synonyms:TEKTB1|TEKTIN-T|h-tektin-t
- Antigen Name:Tektin 1
- ELISA:Yes
- Storage Temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



