Anti-KDM5B Rabbit Polyclonal Antibody
Catalog # 89521-042
Supplier: Abgent
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Lysine (K)-Specific Demethylase 5B
- Antigen Symbol:KDM5B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Mouse
- Western Blot:Yes
- Size:400 µL
- Environmentally Preferable:
- Epitope:N-Terminal
- Gene ID:10765
- Antigen Synonyms:PLU1|JARID1B|CT31|RBP2-H1|PUT1|RBBP2H1A|PPP1R98|PLU-1
- Amino Acid Number:192 to 224
- Storage Buffer:PBS with 0.09% (W/V) sodium azide, pH7.4
- Molecular Weight:175.658 kDa
- Sequence:EYKPHDIPQRQSVQPSETC
- Storage Temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.5mg/ml
- Shipping Temperature:4 °C
- Immunogen:This JARID1B antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 192-224 amino acids from the N-terminal region of human JARID1B.
- Purification:Protein A & Peptide affinity purification
- Cat. No.:89521-042
- Supplier no.:AP14476A
Specifications
About this item
Western Blot: 1:1000
Type: Primary
Antigen: KDM5B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Mouse