About this item
Purified anti-mouse TSLP [28F12]; Reactivity: Mouse; Apps: ELISA Capture, Neut; Size: 500 μg
Each lot of this antibody is quality control tested by ELISA assay. For ELISA capture applications, a concentration range of 1-4 μg/ml is recommended. To obtain a linear standard curve, serial dilutions of TSLP recombinant protein ranging from 2000 to 31.3 pg/ml are recommended for each ELISA plate. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: TSLP
Clonality: Monoclonal
Clone: 28F12
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 10761-980
- 10761-978
- Antigen Symbol:
- TSLP
- TSLP
- Antigen Synonyms:
- thymic stromal derived lymphopoietin
- thymic stromal derived lymphopoietin
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- 28F12
- 28F12
- Reactivity:
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Gene ID:
- 53603
- 53603
- ELISA:
- Yes
- Yes
- Gel Shift:
- Yes
- Yes
- Neutralisation:
- Yes
- Yes
- Immunogen:
- Thymus stromal lymphopoietin
- Thymus stromal lymphopoietin
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Purification:
- The antibody was purified by affinity chromatography.
- The antibody was purified by affinity chromatography.
- Concentration:
- 0.50 mg/mL
- 0.50 mg/mL
- Storage Temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C.
- The antibody solution should be stored undiluted between 2 °C and 8 °C.
- Shipping Temperature:
- Ambient RT
- Ambient RT