Order Entry
United States
ContactUsLinkComponent
Anti-RAB20 Rabbit Polyclonal Antibody
Anti-RAB20 Rabbit Polyclonal Antibody
Catalog # 89517-928
Supplier:  Abgent
Anti-RAB20 Rabbit Polyclonal Antibody
Catalog # 89517-928
Supplier:  Abgent
Supplier Number:  AP17593B
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Ras-related protein Rab-20
  • Antigen Symbol:
    RAB20
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    400 µL
  • Environmentally Preferable:
  • Epitope:
    C-Terminal
  • Gene ID:
    55647
  • Amino Acid Number:
    202 to 229
  • Storage Buffer:
    PBS with 0.09% (W/V) sodium azide, pH7.4
  • Molecular Weight:
    26.277 kDa
  • Sequence:
    VDISSHKPPKRTRC
  • Storage Temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.5mg/ml
  • Shipping Temperature:
    4 °C
  • Immunogen:
    This RAB20 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 202-229 amino acids from the C-terminal region of human RAB20.
  • Purification:
    Protein A & Peptide affinity purification
  • Cat. No.:
    89517-928
  • Supplier no.:
    AP17593B

Specifications

About this item

Western Blot: 1:1000

Type: Primary
Antigen: RAB20
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human