About this item
Purified anti-human SSEA-5 [8E11]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green; Apps: FC, IHC, IP; Size: 200 μg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: SSEA-5
Clonality: Monoclonal
Clone: 8E11
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 10759-596
- 10759-594
- Antigen Symbol:
- SSEA-5
- SSEA-5
- Antigen Synonyms:
- SSEA5|Stage-Specific Embryonic Antigen-5 (SSEA5)
- SSEA5|Stage-Specific Embryonic Antigen-5 (SSEA5)
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- 8E11
- 8E11
- Reactivity:
- African green monkey, Human
- African green monkey, Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Immunogen:
- Human embryonic stem cells
- Human embryonic stem cells
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Purification:
- The antibody was purified by affinity chromatography.
- The antibody was purified by affinity chromatography.
- Concentration:
- 0.50 mg/mL
- 0.50 mg/mL
- Storage Temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C.
- The antibody solution should be stored undiluted between 2 °C and 8 °C.
- Shipping Temperature:
- Ambient RT
- Ambient RT