Ultra-LEAF™ Purified anti-mouse/human LAP (TGF-β1) [TW7-28G11]; Isotype: Mouse IgG2b, κ; Reactivity: Mouse, Human; Apps: FC, WB, IP, Neut; Size: 1 mg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume or 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: LAP
Clonality: Monoclonal
Clone: TW7-28G11
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Mouse;Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 10756-552
- 10759-206
- Antigen Symbol:
- LAP
- LAP
- Antigen Synonyms:
- Latency Associated Peptide (LAP)|TGFB|DPD1|TGF-beta1|TGF-beta|TGFB1
- Latency Associated Peptide (LAP)|TGFB|DPD1|TGF-beta1|TGF-beta|TGFB1
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- TW7-28G11
- TW7-28G11
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- 21803
- 21803
- Isotype:
- IgG2b kappa
- IgG2b kappa
- Western Blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Neutralisation:
- Yes
- Yes
- Immunogen:
- Mouse Tgfb1â€transduced P3U1 cells
- Mouse Tgfb1â€transduced P3U1 cells
- Form:
- 0.2 µm filtered in phosphate-buffered solution, pH 7.2, containing no preservative. Endotoxin level is <0.01 EU/µg of the protein (<0.001 ng/µg of the protein) as determined by the LAL test.
- 0.2 µm filtered in phosphate-buffered solution, pH 7.2, containing no preservative. Endotoxin level is <0.01 EU/µg of the protein (<0.001 ng/µg of the protein) as determined by the LAL test.
- Purification:
- The Ultra-LEAF™ (Low Endotoxin, Azide-Free) antibody was purified by affinity chromatography.
- The Ultra-LEAF™ (Low Endotoxin, Azide-Free) antibody was purified by affinity chromatography.
- Concentration:
- 1.00 mg/mL
- 1.00 mg/mL
- Storage Temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C. This Ultra-LEAF™ solution contains no preservative; handle under aseptic conditions.
- The antibody solution should be stored undiluted between 2 °C and 8 °C. This Ultra-LEAF™ solution contains no preservative; handle under aseptic conditions.
- Shipping Temperature:
- Ambient RT
- Ambient RT