Order Entry
United States
ContactUsLinkComponent
 
Anti-CD1d Mouse Monoclonal Antibody (Brilliant Violet® 510) [clone: 51.1]
  10758-968
 :  Biolegend
Anti-CD1d Mouse Monoclonal Antibody (Brilliant Violet® 510) [clone: 51.1]
  10758-968
 :  Biolegend
 :  350314
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Symbol:
    CD1d
  • Clonality:
    Monoclonal
  • Clone:
    51.1
  • Conjugation:
    Brilliant Violet® 510
  • Host:
    Mouse
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Cynomolgus monkey,
    Human
  • Size:
    100 tests
  • Environmentally Preferable:
  • Form:
    Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
  • Gene ID:
    912
  • Antigen Synonyms:
    R3G1
  • Storage Temperature:
    The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • Shipping Temperature:
    Ambient RT
  • Immunogen:
    Human CD1d-Fc fusion
  • Purification:
    The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 510™ under optimal conditions. The solution is free of unconjugated Brilliant Violet 510™ and unconjugated antibody.
  • Cat. No.:
    10758-968
  • Supplier no.:
    350314

 

 

Brilliant Violet 510™ anti-human CD1d [51.1]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Cynomolgus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD1d
Clonality: Monoclonal
Clone: 51.1
Conjugation: Brilliant Violet® 510
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human;Cynomolgus