Order Entry
United States
ContactUsLinkComponent
Anti-OR4D11 Rabbit Polyclonal Antibody
Anti-OR4D11 Rabbit Polyclonal Antibody
Catalog # 89519-932
Supplier:  Abgent
Anti-OR4D11 Rabbit Polyclonal Antibody
Catalog # 89519-932
Supplier:  Abgent
Supplier Number:  AP16382B
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Olfactory Receptor, Family 4, Subfamily D Member 11
  • Antigen Symbol:
    OR4D11
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    400 µL
  • Environmentally Preferable:
  • Epitope:
    C-Terminal
  • Gene ID:
    219986
  • Antigen Synonyms:
    OR4D11P
  • Amino Acid Number:
    284 to 311
  • Storage Buffer:
    PBS with 0.09% (W/V) sodium azide, pH7.4
  • Molecular Weight:
    34.969 kDa
  • Sequence:
    CRRLKRRLVPSERE
  • Storage Temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.5mg/ml
  • Shipping Temperature:
    4 °C
  • Immunogen:
    This OR4D11 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 284-311 amino acids from the C-terminal region of human OR4D11.
  • Purification:
    Protein A & Peptide affinity purification
  • Cat. No.:
    89519-932
  • Supplier no.:
    AP16382B

Specifications

About this item

Western Blot: 1:1000

Type: Primary
Antigen: OR4D11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human