Order Entry
United States
ContactUsLinkComponent
Anti-MASTL Rabbit Polyclonal Antibody
Anti-MASTL Rabbit Polyclonal Antibody
  89521-564
 :  Abgent
Anti-MASTL Rabbit Polyclonal Antibody
  89521-564
 :  Abgent
 :  AP14289C
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Microtubule Associated Serine/Threonine Kinase Like
  • Antigen Symbol:
    MASTL
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Mouse
  • Western Blot:
    Yes
  • Size:
    400 µL
  • Environmentally Preferable:
  • Epitope:
    Center
  • Gene ID:
    67121
  • Antigen Synonyms:
    hGWL|GWL|MAST-L|THC2|GW|GREATWALL
  • Amino Acid Number:
    561 to 589
  • Storage Buffer:
    PBS with 0.09% (W/V) sodium azide, pH7.4
  • Molecular Weight:
    95.975 kDa
  • Sequence:
    ALEPDKSIRDYSFEC
  • Storage Temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.5mg/ml
  • Shipping Temperature:
    4 °C
  • Immunogen:
    This Mouse Mastl antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 561-589 amino acids from the Central region of mouse Mastl.
  • Purification:
    Protein A & Peptide affinity purification
  • Cat. No.:
    89521-564
  • Supplier no.:
    AP14289C

 

 

Western Blot: 1:1000

Type: Primary
Antigen: MASTL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Mouse