About this item
PE/Dazzle™ 594 anti-human CD16 [3G8]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Capuchin Monkey, Chimpanzee, Cynomolgus, Marmoset, Pigtailed Macaque, Rhesus, Sooty Mangabey, Squirrel Monkey; Apps: FC; Size: 100 tests
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: CD16
Clonality: Monoclonal
Clone: 3G8
Conjugation: PE (Phycoerythrin)/Dazzle 594
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Capuchin Monkey;Chimpanzee;Cynomolgus;Marmoset;Pigtailed Macaque;Rhesus;Sooty Mangabey;Squirrel Monkey
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 10759-444
- 10759-442
- Antigen Symbol:
- CD16
- CD16
- Antigen Synonyms:
- FCGR3A|FcγRIII
- FCGR3A|FcγRIII
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)/Dazzle 594
- PE (Phycoerythrin)/Dazzle 594
- Clone:
- 3G8
- 3G8
- Reactivity:
- Marmoset, Baboon, Capuchin monkey, African green monkey, Cynomolgus monkey, Sooty mangabey, Rhesus monkey, Pigtailed macaque, Chimpanzee, Human, Squirrel monkey
- Marmoset, Baboon, Capuchin monkey, African green monkey, Cynomolgus monkey, Sooty mangabey, Rhesus monkey, Pigtailed macaque, Chimpanzee, Human, Squirrel monkey
- Host:
- Mouse
- Mouse
- Gene ID:
- 2214
- 2214
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Immunogen:
- Human PMN cells
- Human PMN cells
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Purification:
- The antibody was purified by affinity chromatography and conjugated with PE/Dazzle™ 594 under optimal conditions. The solution is free of unconjugated PE/Dazzle™ 594 and unconjugated antibody.
- The antibody was purified by affinity chromatography and conjugated with PE/Dazzle™ 594 under optimal conditions. The solution is free of unconjugated PE/Dazzle™ 594 and unconjugated antibody.
- Storage Temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping Temperature:
- Ambient RT
- Ambient RT