Order Entry
United States
ContactUsLinkComponent
 
Anti-CT19 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: B-D13]
  102995-462
Anti-CT19 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: B-D13]
  102995-462
 :  CDM021
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Name:
    IL-13R alpha 2
  • Antigen Symbol:
    CT19
  • Clonality:
    Monoclonal
  • Clone:
    B-D13
  • Conjugation:
    PE (Phycoerythrin)
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Size:
    200 µg
  • Environmentally Preferable:
  • Antigen Synonyms:
    IL-13R subunit alpha-2|testis antigen 19|CD213A2|IL-13R|interleukin 13 receptor alpha 2 chain|IL-13R-alpha-2|cancer|IL13BP|IL-13 receptor subunit alpha-2|interleukin-13-binding protein|interleukin 13 binding protein|IL-13RA2
  • Cat. No.:
    102995-462
  • Supplier no.:
    CDM021

 

 

Flow Cytometry: Use 10 µl to label 1x10E6 cells or 100 µl of whole blood.

Type: Primary
Antigen: CT19
Clonality: Monoclonal
Clone: B-D13
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity: