ContactUsLinkComponent
Perforated Instrument Trays, Sklar®
Perforated Instrument Trays, Sklar®
Catalog # 10193-432
Supplier:  Sklar
undefined
Perforated Instrument Trays, Sklar®
Catalog # 10193-432
Supplier:  Sklar
Supplier Number:  10-1894
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Instrument Tray, Perforated Bottom
  • Dimensions:
    16.5×10×2.5"
  • Material:
    Stainless steel
  • Environmentally Preferable:
  • Cat. No.:
    10193-432
  • Supplier no.:
    10-1894
Specifications
About this item
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
Perforated Trays are used for sterilizing and transporting instruments for surgery. Available in a variety of sizes, they feature a seamless design to prevent bacterial growth. Perforations also prevent moisture build-up. Dome or flat lids are also available.