ContactUsLinkComponent
Digital Sensor Cable
Digital Sensor Cable
Catalog # 470219-300
undefined
Digital Sensor Cable
Catalog # 470219-300
Supplier Number:  MDC-BTD
Specifications
  • Description:
    Digital sensor cable
  • Environmentally Preferable:
  • Cat. No.:
    470219-300
  • Supplier no.:
    MDC-BTD
Specifications
About this item
This cable connects Vernier digital sensors to the digital (DIG) port on a Vernier interface.
Caution: Not compatible with the Photogate.