ContactUsLinkComponent
LabQuest Viewer Software
LabQuest Viewer Software
Catalog # 470219-296
undefined
LabQuest Viewer Software
Catalog # 470219-296
Supplier Number:  LQ-VIEW
Specifications
  • Description:
    Lab Quest Viewer software
  • Environmentally Preferable:
  • Cat. No.:
    470219-296
  • Supplier no.:
    LQ-VIEW
Specifications
About this item
LabQuest Viewer is computer software that displays and controls the screen of a LabQuest using a computer. When used in conjunction with a projector, this software can be used to demonstrate the functions of LabQuest and to enhance group presentations. This software can also be used to monitor student use of LabQuest devices in the lab, view and control a LabQuest remotely, and easily capture LabQuest screen images for use in lab instructions or assessment questions.
Use LabQuest Viewer in the classroom to:
View and control one or more LabQuest devices from a computer
Use with a projector or interactive white board for class demos and student presentations
Display live images of all LabQuests in your lab to monitor student progress throughout the lab
Select up to four LabQuest devices for side-by-side comparison of experimental results from different lab groups
Capture and share screenshots of the LabQuest screen for use in lab instructions and assessment questions
Set customizable permissions that permit viewing without control, or password-protect a connection
Install LabQuest Viewer on any computer in your school or college department with the included site license
System Requirements:
Windows
Windows 7 SP1, Windows 8, Windows 8.1, and provisional support for Windows 10*
* Tested on a pre-released version of Windows 10; full support in a future release of LabQuest Viewer
Macintosh
OS X 10.8, 10.9 and 10.10
LabQuest Compatibility:
LabQuest 2 (Wi-Fi or USB connection)
LabQuest with Wi-Fi USB Adapter (Wi-Fi only)