Order Entry
United States
ContactUsLinkComponent
Reveal® G4 Rapid HIV-1/2 Antibody Test, Point-of-Care (POC), MedMira
Reveal® G4 Rapid HIV-1/2 Antibody Test, Point-of-Care (POC), MedMira
Catalog # 10829-302
Supplier:  MEDMIRA
CAS Number:  
undefined
Reveal® G4 Rapid HIV-1/2 Antibody Test, Point-of-Care (POC), MedMira
Catalog # 10829-302
Supplier:  MEDMIRA
Supplier Number:  815311007583
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Reveal® G4 Rapid HIV-1/2 Antibody Test, Point-of-Care (POC)
  • Environmentally Preferable:
  • Cat. No.:
    10829-302
  • Supplier no.:
    815311007583

Specifications

About this item

Reveal® G4 Rapid HIV-1/2 Antibody Test uses MedMira Rapid Vertical Flow Technology to deliver instant results in three easy steps.

Reveal® G4 POC enables healthcare professionals at point-of-care settings to test easily for HIV, deliver clear results, and begin treatment immediately if needed. The test features a built-in reagent control line, works with fingerstick whole blood specimens, and requires no additional equipment.
HIV-1 sensitivity: 99.64% Fingerstick Whole Blood
HIV-2 sensitivity: 100%.
Specificity: 99.71% Fingerstick Whole Blood
Test kits can be stored for 12 months at 2 to 30°C (35 to 85°F).

Reveal® G4 is a solution for fast and easy testing in POC settings including:
Public health programs
Voluntary testing and counseling centers
Pharmacies and physician offices
Convenience care clinics
Emergency and first responders

HIV-1/2 Antibody Test Controls are designed for use with Reveal® G4 Rapid HIV-1/2 Antibody Test in customer quality assurance practices. Each set of test controls contains 1 HIV-1 Positive, 1 HIV-2 Positive and 1 HIV-1/2 Negative test control. Each vial is sufficient to perform five Reveal® G4 tests. HIV-1/2 Antibody Test Controls have a shelf life of 12 months, and are stored at room temperature until ready for use.

This simple test provides instant results for either reactive or non-reactive specimens 5 times faster than any lateral flow test. Primary applications for Reveal® G4 include occupational exposures, preventing mother-to-child transmission, and emergency departments.

Certifications: FDA Approved.
Ordering information: Each box of Reveal® G4 POC includes 20 individual test kits. Other formats of Reveal® G4 are available for testing venipuncture whole blood, serum, and plasma specimens. Test contols are available as an accessory. Each set of HIV-1/2 Antibody Test Controls contains1 HIV-1 Positive, 1 HIV-2 Positive and 1 HIV-1/2 Negative test control, each sufficient to run five Reveal® G4 tests.

Controls

Product Information