To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Neuron-specific beta-III Tubulin
- Clonality:Monoclonal
- Clone:TuJ-1
- Conjugation:NL® 637
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Multiple
- Size:500 µL
- Environmentally Preferable:
- Form:Supplied in a saline solution containing BSA and Sodium Azide.
- Antigen Synonyms:Tubulin Beta 3|Tubulin beta-4
- Storage Temperature:Store the unopened product at 2 - 8 °C. Do not use past expiration date. Protect from light.
- Immunogen:Rat brain-derived microtubules
- Purification:Protein A or G purified from hybridoma culture supernatant
- Cat. No.:220021-957
- Supplier no.:NL1195V
Type: Primary
Antigen Symbol:
Antigen Name: Neuron-specific beta-III Tubulin
Antigen Synonyms: Tubulin Beta 3, Tubulin beta-4
Clonality: Monoclonal
Clone: TuJ-1
Conjugation: NL637
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Multiple