Anti-PSMB9 Mouse Monoclonal Antibody [clone: 792520]
102887-320
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Proteasome 20S Beta 9
- Antigen Symbol:PSMB9
- Clonality:Monoclonal
- Clone:792520
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Form:Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
- Gene ID:5698
- Antigen Synonyms:macropain) subunit|proteasome-related gene 2|low molecular mass protein 2|9 (large multifunctional peptidase 2)|beta type|macropain chain 7|proteasome catalytic subunit 1i|proteasome beta 9 subunit|proteasome (prosome|RING12|MGC70470|LMP2|proteasome chain 7|beta1i|PSMB6i|proteasome subunit beta 6i|multicatalytic endopeptidase complex chain 7|PSMB9
- Amino Acid Number:Thr21 to Glu219
- Storage Temperature:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under s
- Shipping Temperature:The product is shipped at ambient temperature. Upon receipt, store it immediately at the temperature recommended below. *Small pack size (SP) is shipped with polar packs. Upon receipt, store it immediately at -20 to -70 °C
- Immunogen:E. coli -derived recombinant human LMP2/PSMB9 Thr21-Glu219; Accession # P28065
- Purification:Protein A or G purified from hybridoma culture supernatant
- Cat. No.:102887-320
- Supplier no.:MAB7709
Type: Primary
Antigen: PSMB9
Clonality: Monoclonal
Clone: 792520
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human