Anti-FZD8 Rat Monoclonal Antibody [clone: 163842]
Type: Primary
Antigen Symbol: FZD8
Antigen Name: frizzled homolog 8 (Drosophila)
Antigen Synonyms: Fz8
Clonality: Monoclonal
Clone: 163842
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 220018-818
- 220018-826
- Antigen Symbol:
- FZD8
- FZD8
- Antigen Name:
- Frizzled 8
- Frizzled 8
- Antigen Synonyms:
- hFZ8|FZ-8
- hFZ8|FZ-8
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 163842
- 163842
- Reactivity:
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Gene ID:
- 14370
- 14370
- Isotype:
- IgG2a
- IgG2a
- Western Blot:
- Yes
- Yes
- Immunogen:
- Mouse myeloma cell line NS0-derived recombinant mouse Frizzled-8 Ala28-Pro173 (predicted); Accession # Q61091
- Mouse myeloma cell line NS0-derived recombinant mouse Frizzled-8 Ala28-Pro173 (predicted); Accession # Q61091
- Amino Acid Number:
- Ala28 to Pro173 (predicted)
- Ala28 to Pro173 (predicted)
- Form:
- Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
- Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
- Purification:
- Protein A or G purified from hybridoma culture supernatant
- Protein A or G purified from hybridoma culture supernatant
- Storage Temperature:
- Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under s
- Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under s
- Shipping Temperature:
- The product is shipped at ambient temperature. Upon receipt, store it immediately at the temperature recommended below. *Small pack size (SP) is shipped with polar packs. Upon receipt, store it immediately at -20 to -70 °C
- The product is shipped at ambient temperature. Upon receipt, store it immediately at the temperature recommended below. *Small pack size (SP) is shipped with polar packs. Upon receipt, store it immediately at -20 to -70 °C
Frequently Bought Together