Order Entry
United States
ContactUsLinkComponent
Anti-S1PR1 Rat Monoclonal Antibody (Biotin) [clone: 713412]
Anti-S1PR1 Rat Monoclonal Antibody (Biotin) [clone: 713412]
Catalog # 102883-552
Supplier:  R&D Systems
Anti-S1PR1 Rat Monoclonal Antibody (Biotin) [clone: 713412]
Catalog # 102883-552
Supplier:  R&D Systems
Supplier Number:  FAB7089B
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Sphingosine-1-phosphate Receptor 1
  • Antigen Symbol:
    S1PR1
  • Clonality:
    Monoclonal
  • Clone:
    713412
  • Conjugation:
    Biotin
  • Flow Cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG2a
  • Reactivity:
    Mouse
  • Size:
    100 Tests
  • Environmentally Preferable:
  • Form:
    Supplied in a saline solution containing BSA and Sodium Azide.
  • Gene ID:
    13609
  • Antigen Synonyms:
    CD363|CHEDG1|Sphingosine 1-phosphate receptor 1|S1P receptor 1|ECGF1|D1S3362|EDG-1|Endothelial differentiation G-protein coupled receptor 1|EDG1|S1P1
  • Storage Temperature:
    Do not freeze.12 months from date of receipt, 2 to 8 °C as supplied.
  • Shipping Temperature:
    The product is shipped with polar packs. Upon receipt, store it immediately at the temperature recommended below.
  • Immunogen:
    Mouse S1P1/EDG‑1 synthetic peptide (T4-H28); Accession # O08530
  • Purification:
    Protein A or G purified from hybridoma culture supernatant
  • Cat. No.:
    102883-552
  • Supplier no.:
    FAB7089B

Specifications

About this item

Type: Primary
Antigen: S1PR1
Clonality: Monoclonal
Clone: 713412
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse