Anti-KIR2DL1 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 143211]
Supplier: R&D Systems
Certificates
About this item
Type: Primary
Antigen: KIR2DL1
Clonality: Monoclonal
Clone: 143211
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 102881-512
- 102881-510
- Antigen Symbol:
- KIR2DL1
- KIR2DL1
- Antigen Name:
- Killer cell immunoglobulin-like Receptor two Domains long cytoplasmic tail 1
- Killer cell immunoglobulin-like Receptor two Domains long cytoplasmic tail 1
- Antigen Synonyms:
- NKAT-1|p58.1|NKAT1|CD158A|KIR-K64|NKAT|KIR221
- NKAT-1|p58.1|NKAT1|CD158A|KIR-K64|NKAT|KIR221
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PerCP (Peridinin-Chlorophyll Protein Complex)
- PerCP (Peridinin-Chlorophyll Protein Complex)
- Clone:
- 143211
- 143211
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- BaF3 mouse pro-B cell line transfected with human KIR2DL1/KIR2DS5; Accession # P43626
- BaF3 mouse pro-B cell line transfected with human KIR2DL1/KIR2DS5; Accession # P43626
- Form:
- Supplied in a saline solution containing BSA and Sodium Azide.
- Supplied in a saline solution containing BSA and Sodium Azide.
- Purification:
- Protein A or G purified from hybridoma culture supernatant
- Protein A or G purified from hybridoma culture supernatant
- Storage Temperature:
- Protect from light. Do not freeze.12 months from date of receipt, 2 to 8 °C as supplied.
- Protect from light. Do not freeze.12 months from date of receipt, 2 to 8 °C as supplied.
- Shipping Temperature:
- The product is shipped with polar packs. Upon receipt, store it immediately at the temperature recommended below.
- The product is shipped with polar packs. Upon receipt, store it immediately at the temperature recommended below.
Specifications
Frequently Bought Together