Anti-CCL27A Rat Monoclonal Antibody (Biotin) [clone: 68706]
220014-653
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Chemokine (C-C motif) ligand 27A
- Antigen Symbol:CCL27A
- Clonality:Monoclonal
- Clone:68706
- Conjugation:Biotin
- Host:Rat
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Mouse
- Western Blot:Yes
- Size:250 µg
- Environmentally Preferable:
- Form:Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
- Gene ID:20301
- Antigen Synonyms:PESKY|ILC|Scya27|CTACK|ESkine|AW558992|Ccl27|CTAK|ALP|Scya27a
- Amino Acid Number:Leu26 to Asn120
- Storage Temperature:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under s
- Shipping Temperature:The product is shipped at ambient temperature. Upon receipt, store it immediately at the temperature recommended below.
- Immunogen:E. coli -derived recombinant human CCL27/CTACK Leu26-Asn120; Accession # NP_035466
- Purification:Protein A or G purified from hybridoma culture supernatant
- Cat. No.:220014-653
- Supplier no.:BAM7251
Type: Primary
Antigen: CCL27A
Clonality: Monoclonal
Clone: 68706
Conjugation: Biotin
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse