To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:heparin-binding EGF-like growth factor
- Antigen Symbol:HBEGF
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:250 µg
- Environmentally Preferable:
- Neutralisation:Yes
- Form:Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
- Gene ID:1839
- Antigen Synonyms:DTR|HEGFL|DTS|DTSF
- Amino Acid Number:Asp63 to Leu148
- Storage Temperature:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under s
- Shipping Temperature:The product is shipped at ambient temperature. Upon receipt, store it immediately at the temperature recommended below. *Small pack size (SP) is shipped with polar packs. Upon receipt, store it immediately at -20 to -70 °C
- Immunogen:S. frugiperda insect ovarian cell line Sf 21-derived recombinant human HB-EGF (R&D Systems, Catalog # 259-HE) Asp63-Leu148; Accession # Q99075
- Purification:Antigen Affinity-purified
- Cat. No.:220010-623
- Supplier no.:AF-259-NA
Specifications
About this item
Type: Primary
Antigen Symbol: HBEGF
Antigen Name: heparin-binding EGF-like growth factor
Antigen Synonyms: DTR, DTS, DTSF, HEGFL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human