Order Entry
United States
ContactUsLinkComponent
MRO Plus™ Heavy Roll, Brady
MRO Plus™ Heavy Roll, Brady
Catalog # 10809-782
Supplier:  Brady Worldwide
undefined
MRO Plus™ Heavy Roll, Brady
Catalog # 10809-782
Supplier:  Brady Worldwide
Supplier Number:  107727

Specifications

  • Description:
    Roll
  • Environmentally Preferable:
  • Capacity:
    24 gal.
  • Dimensions:
    15W" x 150L'
  • Cat. No.:
    10809-782
  • Supplier no.:
    MRO15P

Specifications

About this item

The MRO Plus™ Heavy Roll absorbs all industrial liquids (water, petroleum-based and chemical-based fluids)

3-ply construction makes it stronger, more durable and reduces lint. Gray color masks drips and spills, dimpled, perfed so you can take only what you need and reduce waste, heavy weight roll. Perfed every 18". For use with oil and water-based fluids. Polypropylene material. Gray color.