ContactUsLinkComponent
Pierce™ Endoproteinase Lys-C Protease, MS Grade, Thermo Scientific
Pierce™ Endoproteinase Lys-C Protease, MS Grade, Thermo Scientific
Catalog # PI90051
Supplier:  Invitrogen
CAS Number:  
undefined
Pierce™ Endoproteinase Lys-C Protease, MS Grade, Thermo Scientific
Catalog # PI90051
Supplier:  Invitrogen
Supplier Number:  90051
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Endoproteinase Lys-C protease, MS grade
  • Size:
    1 x 20 µg
  • Environmentally Preferable:
  • Cat. No.:
    PI90051
  • Supplier no.:
    90051
Specifications
About this item
Endoproteinase Lys-C protease is a highly purified native endoproteinase validated for maximum activity and stability in proteomic applications.
  • Can be used in tandem with trypsin to decrease tryptic missed cleavages thus enhancing digestion
  • Better protein characterization results from overlapping peptides with complementary chromatographic, ionisation and fragmentation properties thereby increasing sequence coverage
  • Effective enzyme activity under highly denaturing conditions
  • C-terminal lysine cleavage specificity of 90% for a complex protein sample
  • Stable since it is provided in a lyophilized format
This Lys-C is a mass spectrometry (MS)-grade serine protease isolated from Lysobacter enzymogenes
Applications include improved sequence coverage of protein digests, de novo sequencing, epigenetic studies and in-gel and in-solution digestion of proteins.
Lys-C has high activity and specificity for lysine residues resulting in larger peptides and less sample complexity than trypsin (i.e. fewer peptides). Unlike trypsin, Lys-C can cleave lysines followed by prolines, making it ideal for sequential protein digestion followed by trypsin to decrease missed cleavages. These unique Lys-C properties ensure high digestion efficiency when used alone or followed by tryptic digestion.
Caution: For Research Use Only. Not for use in diagnostic procedures.