Specifications
- Description:Nylon membrane, sheets, pore size 0.45 µm
- Dimensions:8 x 12 cm
- Size:25 Sheets
- Environmentally Preferable:
- Cat. No.:PI77016
- Supplier no.:77016
Specifications
About this item
Thermo Scientific Biodyne B Nylon Membranes are sheets of positively charged nylon that provide excellent binding characteristics for nucleic acid transfer, blotting and detection methods.
- Versatile and durable—achieve strong DNA binding by baking or UV-crosslinking without damaging the membrane
- Easy to use—pre-cut 8 x 12 cm sheets are the ideal size for standard mini-gel transfers and microplate dot-blots
- High signal-to-noise ratio—lower background than nitrocellulose and most other nylon transfer membranes
Biodyne B Membrane contains exclusively positive charges, providing the highest possible binding capacity for negatively charged molecules like DNA and RNA
Properties of Biodyne B Nylon Membrane:
Composition: positively charged Nylon 66
Surface: strong cationic quaternary ammonium groups provide positive zeta potential, even to pH 10
Properties: excellent for DNA, RNA and other negatively charged molecules
Applications: Southern blotting, northern blotting, DNA and RNA methods, including gel shift assays with Thermo Scientific LightShift Chemiluminescent EMSA Kits.
The membrane is resistant to heat and solvents and will not shrink, crack or tear.