Specifications
- Description:Adson-Beckman Retractor
- Length:31.8 cm (12½")
- Material:Stainless steel
- Environmentally Preferable:
- Tip configuration:4x4 Prongs Sharp
- Cat. No.:10197-690
- Supplier no.:57-1322
Specifications
About this item
The Adson-Beckman Retractor is a finger ring, self-retaining retractor used in neurosurgical procedures, most commonly related to the spine
This Adson-Beckman Retractor features hinged arms with pronged tips that are used to hold back muscle and tissue. The arms move both backwards and forwards and the prongs may be blunt or sharp.