To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Glucosidase alpha; neutral C
- Antigen Symbol:GANC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Cross Adsorption:No
- Form:liquid
- Gene ID:2595
- Antigen Synonyms:Neutral alpha-glucosidase C|MGC138256|Neutral alphaglucosidase C.|GANC_HUMAN|Neutral alpha glucosidase C|Glucosidase alpha neutral C|Ganc
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
- Storage Temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10295-662
- Supplier no.:BS-13279R
A key enzyme in glycogen degradation and metabolism, GANC (glucosidase, α neutral C) is a 914 amino acid protein with α-glucosidase activity that belongs to the glycosyl hydrolase 31 family and hydrolyzes non-reducing, terminal 1,4-linked α-D-glucose residues and releases α-D-glucose. The gene encoding GANC maps to human chromosome 15q15.1, a region associated with susceptibility to non-insulin-dependent (type 2) diabetes mellitus, a disease characterized by high blood glucose levels. Human chromosome 15 houses over 700 genes and comprises nearly 3% of the human genome. Angelman syndrome, Prader-Willi syndrome, Tay-Sachs disease and Marfan syndrome are all associated with defects in chromosome 15-localized genes.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200
Type: Primary
Antigen: GANC/Neutral alphaglucosidase C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat