To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:APCL/APC2
- Antigen Symbol:APCL/APC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Cross Adsorption:No
- Form:liquid
- Gene ID:10297
- Antigen Synonyms:Adenomatous polyposis coli protein-like|APC2_HUMAN.|APCL|Adenomatous polyposis coli protein 2
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
- Storage Temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10284-050
- Supplier no.:BS-12485R
Specifications
About this item
The adenomatous polyposis syndromes, familial adenomatous polyposis (FAP) and Gardner’s syndrome (GS), are characterized by numerous adenomatous polyps throughout the entire colon. These polyps invariably progress to colon cancer in addition to other extracolonic manifestations. The cloning of the APC gene revealed a ubiquitously expressed protein, 2843 amino acids in length, which is frequently mutated in patients suffering from FAP and GS. APC has been found to be associated with structural components of intracellular junctions. b-catenin and g-catenin (also called plakoglobin) are involved in the regulation of cellular adhesion. APC and E cadherin compete for binding to specific internal regions of both b- and g-catenin. Interactions between cytoskeleton and the APC, E cadherin, b/g catenin complex are mediated by a-catenin.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200
Type: Primary
Antigen: APCL/APC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat