To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Newborn Ovary Homeobox Protein
- Antigen Symbol:NOBOX
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Cross Adsorption:No
- Form:liquid
- Gene ID:135935
- Antigen Synonyms:POF5|TCAG_12042|OG2X|OG-2|OG2
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
- Storage Temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10277-356
- Supplier no.:BS-12273R
Early ovarian folliculogenesis is characterized by the breakdown of germ cell clusters and formation of primordial follicles. The cessation of ovarian function under the age of 40 years results in premature ovarian failure (POF) and is accompanied by amenorrhea, hypoestrogenism and elevated serum gonadotropin concentrations. 1% of all women are affected by POF, and mutations in a few genes, including inhibin? fsh receptor and the LH/choriogonadotropin receptor have been linked to POF. In addition, several germ cell specific genes and downstream transcription factors are thought to play and important role in human oogenesis. NOBOX (newborn ovary homeobox gene), an ooctye-specific homeobox gene, is a critical protein involved in early folliculogenesis. Missense mutations have been shown to cause nonsyndromic ovarian failure by disrupting the NOBOX proteins ability to bind to NOBOX DNA-binding element (NBE), and thereby inhibiting its regulation of Pou5f1 and GDF-9. NOBOX expression in the ovary is oocyte specific, but it shows expression in adult testis and pancreas as well.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200
Type: Primary
Antigen: NOBOX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat