To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:RPGRIP1-like
- Antigen Symbol:RPGRIP1L
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Cross Adsorption:No
- Form:liquid
- Antigen Synonyms:RPGRIP1-like |DKFZp686C0668 |MKS5|CORS3 |KIAA1005 |JBTS7
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
- Storage Temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10268-616
- Supplier no.:BS-11946R
Specifications
About this item
RPGRIP1L is a 1,315 amino acid protein that belongs to the RPGRIP1 family and is thought to function in programmed cell death, craniofacial development and formation of the left-right axis. Existing as two alternatively spliced isoforms that localize to the cytoplasm, cytoskeleton, centrosome and cilium basal body, RPGRIP1L interacts with nephrocystin-4 and is moderately expressed in brain, retina and kidney. Containing two C2 domains, RPGRIP1L is encoded by a gene that maps to human chromosome 16q12.2. Defects in the gene encoding RPGRIP1L are the cause of Joubert syndrome type 7 (JBTS7), COACH syndrome (COACHS) and Meckel syndrome type 5 (MKS5).
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200
Type: Primary
Antigen: RPGRIP1L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat