To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Thymocyte selection-associated high mobility group box 2
- Antigen Symbol:TOX2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Cross Adsorption:No
- Form:liquid
- Gene ID:84969
- Antigen Synonyms:C20orf100|dJ1108D11.2|dJ495O3.1|GCX-1|GCX1
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
- Storage Temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10255-040
- Supplier no.:BS-11402R
GCX-1 is a 488 amino acid transcription activator that is restricted to expression in the hypothalamus, pituitary, ovary, testis and uterus.Human reproduction is controlled by the hypothalamic-pituitary gonadal axis (HPGA) that is laid down early in fetal development. This expression pattern in the HPGA suggests that this nuclear protein is likely related to specific events in reproduction, particularly in the female. GCX-1 contains a HMG-box domain, which is commonly found in proteins that function as intercellular regulators and transcriptional co-regulators, and are found to be involved in important events such as sex determination and in the regulation of T cell differentiation. There are two isoforms of GCX-1 which are produced as a result of alternative splicing events.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200
Type: Primary
Antigen: TOX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat