Order Entry
United States
ContactUsLinkComponent
Anti-Estrogen Receptor alpha + beta Rabbit Polyclonal Antibody
Anti-Estrogen Receptor alpha + beta Rabbit Polyclonal Antibody
Catalog # 10232-596
Supplier:  Bioss
undefined
Anti-Estrogen Receptor alpha + beta Rabbit Polyclonal Antibody
Catalog # 10232-596
Supplier:  Bioss
Supplier Number:  BS-0174R
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Estrogen Receptor alpha + beta
  • Antigen Symbol:
    Estrogen Receptor alpha + beta
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Environmentally Preferable:
  • Cross Adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    2099, 2100
  • Antigen Synonyms:
    Estradiol receptor|Nuclear receptor subfamily 3 group A member 2|Estrogen Receptor alpha/beta|ER|OTTHUMP00000017719|Era|ESR BETA|Estrogen receptor alpha|ER-beta|Estrogen receptor 2 ER beta|ERalpha|Estr|augmented response of|Estradiol Receptor alpha|Estrogen Receptor 2|ESRA|OTTHUMP00000017718|NR3A1|ESR2|Estrogen receptor|Atherosclerosis|susceptibility to|ER-alpha|ER Beta|ESR|ESR1|HDL cholesterol|to hormone replacement|Nuclear receptor subfamily 3 group A member 1|Estrogen Receptor 1|ESRB|included|Erb|Estradiol Receptor beta|NR3A2|Estra|Estrogen resistance|Estrogen receptor 1 alpha|ER Alpha|Estrogen receptor beta 4|Erb2|ESTRB|DKFZp686N23123|ER[a]|ESR1_HUMAN|Myocardial infarction|ER[b]|ERbeta|RNESTROR.
  • Storage Buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
  • Storage Temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping Temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Cat. No.:
    10232-596
  • Supplier no.:
    BS-0174R

Specifications

About this item

The estrogen receptor (ER) is a 66 kDa protein that mediates the actions of estrogens in estrogen responsive tissues. It is a member of a large superfamily of nuclear hormone receptors that function as ligand activated transcription factors. The ER gene consists of more than 140 kb of genomic DNA divided into 8 exons. These translate into a protein with six functionally discrete domains, labeled A through F. A second form of the estrogen receptor, ER beta has recently been described. The ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti estrogen (e.g. tamoxifen) therapy.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: Estrogen Receptor alpha + beta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat