Order Entry
United States
ContactUsLinkComponent
Anti-C18ORF8 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-C18ORF8 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 10494-338
Supplier:  Bioss
Anti-C18ORF8 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 10494-338
Supplier:  Bioss
Supplier Number:  BS-9676R-FITC
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Chromosome 18 Open Reading Frame 8
  • Antigen Symbol:
    C18ORF8
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Size:
    100 µL
  • Environmentally Preferable:
  • Cross Adsorption:
    No
  • Form:
    liquid
  • Antigen Synonyms:
    Colon cancer associated protein Mic1|HsT2591|Chromosome 18 open reading frame 8|Colon cancer-associated protein Mic1|C18orf8|Mic-1|Uncharacterized protein C18orf8.|MIC1_HUMAN|MIC1
  • Storage Buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage Temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping Temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Cat. No.:
    10494-338
  • Supplier no.:
    BS-9676R-FITC

Specifications

About this item

MIC1 is a 657 amino acid protein that contains one MIC1 domain and is encoded by a gene which maps to human chromosome 18. Chromosome 18 houses over 300 protein-coding genes and contains nearly 76 million bases. There are a variety of diseases associated with defects in chromosome 18-localized genes, some of which include Trisomy 18 (also known as Edwards syndrome), Niemann-Pick disease, hereditary hemorrhagic telangiectasia, erythropoietic protoporphyria and follicular lymphomas.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: C18ORF8
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human