To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Tetraspanin 15
- Antigen Symbol:TSPAN15
- Clonality:Polyclonal
- Conjugation:Cy5®
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Form:liquid
- Gene ID:23555
- Antigen Synonyms:Tetraspanin 15|Tetraspanin-15|TSPAN15.|TSN15_HUMAN|Transmembrane 4 superfamily member 15|Tetraspan NET-7|Tspan-15
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10487-874
- Supplier no.:BS-9425R-CY5
Specifications
About this item
NET-7, also known as TSPAN15 (tetraspanin 15) or TM4SF15 (transmembrane 4 superfamily member 15), is a 294 amino acid multi-pass membrane protein that belongs to the transmembrane 4 superfamily, also known as the tetraspanin family. Members of the tetraspanin family are cell-surface proteins that are characterized by the presence of four hydrophobic domains and mediate signal transduction events that play a role in the regulation of cell development, activation, growth, motility, differentiation, and cancer. Considered molecular facilitators, tetraspanin proteins may regulate vesicle fusion and fission.
Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200
Type: Primary
Antigen: TSPAN15
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat