Order Entry
United States
ContactUsLinkComponent
Anti-Transketolase (CT) Rabbit Polyclonal Antibody (Cy5.5®)
Anti-Transketolase (CT) Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # 10397-990
Supplier:  Avantor
Anti-Transketolase (CT) Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # 10397-990
Supplier:  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Transketolase (CT)
  • Antigen Symbol:
    Transketolase (CT)
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy5.5®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Plant
  • Size:
    100 µL
  • Environmentally Preferable:
  • Cross Adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    7086
  • Storage Buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage Temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping Temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Cat. No.:
    10397-990
  • Supplier no.:
    BS-4514R-CY5.5

Specifications

About this item

Transketolase (CT)

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Transketolase (CT)
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Plant