Anti-EDNRB Rabbit Polyclonal Antibody
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
The protein encoded by this gene is a G protein-coupled receptor which activates a phosphatidylinositol-calcium second messenger system. Its ligand, endothelin, consists of a family of three potent vasoactive peptides: ET1, ET2, and ET3. Studies suggest that the multigenic disorder, Hirschsprung disease type 2, is due to mutations in the endothelin receptor type B gene. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq].
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200
Type: Primary
Antigen: EDNRB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Documents
bs-4198R
Datasheet
Specifications
- Size:100 µL
- Cross Adsorption:No
- Clonality:Polyclonal
- Purification:Purified by Protein A
- Form:liquid
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
- Isotype:IgG
- Host:Rabbit
- Concentration:1 μg/μl
- Cat. No.:10401-808
- Reactivity:Human|Rat|Mouse
- Western Blot:Yes
- Shipping Temperature:4°C
- Antibody Type:Primary
- Antigen Symbol:EDNRB
- Gene ID:1910
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen Synonyms:ETB1|ETBR|ET-BR|WS4A|ABCDS|HSCR|ET-B|HSCR2|ETB|ETRB
- Antigen Name:Endothelin receptor type B
- Storage Temperature:Store at –20 °C for 12 months
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



