Order Entry
United States
ContactUsLinkComponent
Anti-CYP11B1 Rabbit Polyclonal Antibody (Cy5.5®)
Anti-CYP11B1 Rabbit Polyclonal Antibody (Cy5.5®)
  10389-546
 :  Bioss
Anti-CYP11B1 Rabbit Polyclonal Antibody (Cy5.5®)
  10389-546
 :  Bioss
 :  BS-3898R-CY5.5
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Cytochrome P450, Family 11, Subfamily B, Polypeptide 1
  • Antigen Symbol:
    CYP11B1
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy5.5®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Size:
    100 µL
  • Environmentally Preferable:
  • Cross Adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    1584
  • Antigen Synonyms:
    CPN1|P450C11|cytochrome P450|cytochrome P-450c11|cytochrome P450 11B1|subfamily XIB (steroid 11-beta-hydroxylase)|FHI|cytochrome p450 XIB1|polypeptide 1|cytochrome P450C11|mitochondrial|steroid 11-beta-monooxygenase|CYP11B|steroid 11-beta-hydroxylase|CYPXIB1
  • Storage Buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage Temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping Temperature:
    4°C
  • Immunogen:
    350-400/503
  • Purification:
    Purified by Protein A
  • Cat. No.:
    10389-546
  • Supplier no.:
    BS-3898R-CY5.5

 

 

Has steroid 11-beta-hydroxylase activity. In addition to this activity, the 18 or 19-hydroxylation of steroids and the aromatization of androstendione to estrone have also been ascribed to cytochrome P45 XIB.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CYP11B1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype:
Reactivity: