To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:SH2 Domain Containing 2A
- Antigen Symbol:SH2D2A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Form:liquid
- Gene ID:9047
- Antigen Synonyms:SH2 domain-containing protein 2A|T cell specific adapter protein TSAd|SH2 domain containing protein 2A|T cell specific adpater protein TSAd|Sh2d2a|T lymphocyte specific adaptor protein|VEGF receptor associated protein|SCAP|SH2 domain protein 2A|SH2 domain containing adapter protein|VEGF receptor-associated protein|VRAP|T cell-specific adapter protein|SH2 domain-containing adapter protein|TSAd|SH22A|T cell specific adapter protein
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
- Storage Temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10395-130
- Supplier no.:BS-3858R
SH2D2A is thought to be a T-cell-specific adapter protein involved in the control of T-cell activation. It may play a role in the CD4-p56-LCK-dependent signal transduction pathway and also in normal and pathological angiogenesis.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200
Type: Primary
Antigen: SH2D2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat