Anti-NPRB Rabbit Polyclonal Antibody
100 µL
Catalog # 10331-842
Supplier: Bioss
Supplier #: BS-2348R
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Receptor for the C-type natriuretic peptide NPPC/CNP hormone. Has guanylate cyclase activity upon binding of its ligand. May play a role in the regulation of skeletal growth.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200
Type: Primary
Antigen: NPRB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Documents
bs-2348R
Datasheet
Specifications
- Size:100 µL
- Cross Adsorption:No
- Clonality:Polyclonal
- Purification:Purified by Protein A
- Form:liquid
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
- Isotype:IgG
- Host:Rabbit
- Concentration:1 μg/μl
- Cat. No.:10331-842
- Reactivity:Human|Rat|Mouse
- Western Blot:Yes
- Shipping Temperature:4°C
- Antibody Type:Primary
- Antigen Symbol:NPRB
- Gene ID:4882
- Immunogen:122-165/1047
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen Synonyms:Guanylate Cyclase B|Atrionatriuretic Peptide Receptor B
- Antigen Name:Natriuretic Peptide Receptor B
- Storage Temperature:Store at –20 °C for 12 months
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



