To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Glutathione Transferase Zeta 1
- Antigen Symbol:GSTZ1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Form:liquid
- Antigen Synonyms:Glutathione S transferase zeta 1|MAAI_HUMAN.|EC 5.2.1.2|GSTZ1-1|EC 2.5.1.18|MAAI
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10298-076
- Supplier no.:BS-13442R-A647
Specifications
About this item
GSTZ1 is a member of the glutathione S transferase (GSTs) super family, encoding multifunctional enzymes important in the detoxification of electrophilic molecules, including carcinogens, mutagens, and several therapeutic drugs, by conjugation with glutathione. This enzyme also plays a significant role in the catabolism of phenylalanine and tyrosine. Several transcript variants of this gene encode multiple protein isoforms. GSTZ1 shows minimal glutathione-conjugating activity with ethacrynic acid and 7-chloro-4-nitrobenz-2-oxa-1,3-diazole and maleylacetoacetate isomerase activity. It has low glutathione peroxidase activity with T butyl and cumene hydroperoxides and is able to catalyze the glutathione dependent oxygenation of dichloroacetic acid to glyoxylic acid. Highest expression in liver followed by kidney, skeletal muscle and brain. Also expressed in melanocytes, synovium, placenta, breast and fetal liver and heart.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: Glutathione Transferase zeta 1/MAAI
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat