To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Glucose Oxidase
- Antigen Symbol:GOX
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Multiple
- Western Blot:Yes
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Form:liquid
- Antigen Synonyms:GOD|Beta D Glucose Oxygen 1 Oxido Reductase|Glucose Oxyhydrase|Glucose oxidase (Precursor)
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10297-634
- Supplier no.:BS-13387R-HRP
Specifications
About this item
Glucose Oxidase is a dimeric enzyme that binds to β-D-glucose and aids in its oxidation into D-glucono-1,5-lactone which then hydrolyzes to gluconic acid. Flavin adenine dinucleotide (FAD) is a cofactor to Glucose Oxidase that acts as the initial electron acceptor and is required for this oxidation to occur. Glucose Oxidase is a natural preservative found in honey, where it reduces atmospheric oxygen into hydrogen peroxide which acts as an antibacterial barrier. Glucose Oxidase is also commonly used in biosensors in which it conveys levels of glucose by keeping track of the number of electrons passed through the enzyme. In this application, Glucose Oxidase is connected to an electrode and the resulting charge is measured.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500
Type: Primary
Antigen: Glucose Oxidase
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Others