To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Glucose-6-phosphatase, Catalytic, 2
- Antigen Symbol:G6PC2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Form:liquid
- Gene ID:57818
- Antigen Synonyms:IGRP
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10297-598
- Supplier no.:BS-13386R-A555
Glucose-6-phosphatase (G6Pase), is a multicomponent enzyme system that hydrolyzes glucose-6-phosphate in the final step of gluconeogenesis and gluconeolysis. G6Pase localizes to the endoplasmic reticulum, and while liver, kidney, and intestine are the only tissues that express the first identified isoform, G6Pase-alpha, a second form, designated G6Pase-∫, contributes to blood glucose homeostasis in a wider range of tissues. Islet-specific G-6-Pase catalytic subunit-related protein (IGRP), a homolog of the catalytic subunit of G6Pase, may play a role in the regulation of islet metabolism and in insulin secretion induced by metabolites. The exact catalytic acivity of IGRP is not defined. Identification of inhibitors of IGRP have potential therapeutic benefits for treatment of type 2 diabetes resulting from insulin secretion defects. Structurally, IGRP has been shown to be a glycoprotein held in the endoplasmic reticulum by nine transmembrane domains, which are then degraded in cells through the proteasome pathway generating MHC class I presented peptides.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: Glucose 6 phosphatase 2/IGRP
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat