To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Fibronectin type III Domain Containing 4
- Antigen Symbol:FNDC4
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Form:liquid
- Gene ID:64838
- Antigen Synonyms:FRCP1
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10293-680
- Supplier no.:BS-13192R-FITC
Specifications
About this item
Fibronectins are multi-domain glycoproteins that bind to a variety of substances including collagen, actin, heparin, DNA, fibrin and fibronectin receptors. They are involved in a diverse array of important functions such as blood coagulation, wound healing, cell adhesion, cell differentiation and migration. FNDC4 (Fibronectin type III domain-containing protein 4), also known as FRCP1 (Fibronectin type III repeat-containing protein 1), is a 234 amino acid membrane protein that contains one fibronectin type-III domain, which serves as a binding site for DNA, heparin or the cell surface. The gene encoding FNDC4 is localized to human chromosome two, which houses over 1,400 genes and comprises nearly 8% of the human genome.
Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200
Type: Primary
Antigen: FNDC4
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat