To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Fructosamine 3 kinase
- Antigen Symbol:FN3K
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Form:liquid
- Gene ID:64122
- Antigen Synonyms:Fnsk|Fructosamine 3 kinase|RP23-293H17.4|2310074G21Rik|FN3K_HUMAN.
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10291-448
- Supplier no.:BS-13189R-CY7
Specifications
About this item
Amines, including those present on proteins, spontaneously react with glucose to make fructosamines in a reaction termed glycation. Fructosamine 3-kinase (FN3K), a 309-amino acid enzyme initially identified in erythrocytes, catalyzes the ATP-dependent phosphorylation of the third carbon on both D- and L-fructosamines, leading to their destabilization and eventually, their removal from the protein. FN3K is a monomer that is ubiquitously expressed in mammalian tissue and phosphorylates both low molecular mass and protein-bound fructosamines which are formed as a result of glycation of glucose with primary amines. FN3K protects proteins from the harmful effects of nonenzymatic glycation, and may also be involved in peptide repair and cell metabolism. FN3KRP (fructosamine-3-kinase-related protein) is a 309 amino acid protein that is expressed in erythrocytes, bone marrow, spleen, brain and kidney and belongs to the fructosamine kinase family. FN3KRP functions to phosphorylate psicoamines and ribulosamines on the third carbon of their sugar moiety, thereby leading to the deglycation of the target amines.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: FN3K
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat