To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:FERM RhoGEF (ARHGEF) and pleckstrin Domain Protein 1
- Antigen Symbol:FARP1
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Form:liquid
- Gene ID:10160
- Antigen Synonyms:FERM RhoGEF ARHGEF and pleckstrin domain protein 1 chondrocyte-derived. PLEKHC2|FERM RhoGEF and pleckstrin domain containing protein 1|CDEP|FARP1_HUMAN.|Chondrocyte derived ezrin like protein
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10293-134
- Supplier no.:BS-13145R-CY5
Specifications
About this item
Aminoacyl-tRNA synthetases consist of a family of enzymes that catalyze the specific aminoacylation of tRNA by their cognate amino acid in the initial step of ribosome-dependent protein biosynthesis. FARSLA, also known as FRSA, CML33, FARSL or PheHA (phenylalanyl-tRNA synthetase, alpha subunit), is a member of the class-II aminoacyl-tRNA synthetase family and is highly expressed in proliferating cells of bone marrow. FARSLA is a cytoplasmic phenylalanine-tRNA synthetase that functions as a heterodimer consisting of a catalytic alpha-subunit and a regulatory beta-subunit. The alpha-subunit is responsible for forming the amino acid binding pocket, mediating the ATP/aminoacyl adenylate binding, and interacts with the acceptor stem of the tRNA. FARSLA functions in a cell cycle-dependent and differentiation-dependent manner.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: FARSLA/CML33
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human