To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Polycomb protein EED
- Antigen Symbol:EED
- Clonality:Polyclonal
- Conjugation:Cy3®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Form:liquid
- Gene ID:8726
- Antigen Synonyms:HEED|WD protein associating with integrin cytoplasmic tails 1|WAIT1|polycomb protein EED
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10289-772
- Supplier no.:BS-13053R-CY3
The transcriptional repressing Polycomb-group (PcG) and transcriptional activating trithorax-group (trxG) genes of Drosophila are part of a cellular memory system responsible for the stable inheritance of gene activity. PcG proteins assemble into multimeric protein complexes, which are involved in maintaining the transcriptional repressive state of genes over successive cell generations. EED (embryonic ectoderm development) is the human homolog of Eed, a murine PcG gene homologous to the Drosophila homeotic gene, extra sex combs. The human EED protein is 99. 5% identical to the mouse EED protein and contains seven WD repeats, which are involved in protein-protein interactions. There are two human EED transcripts that contain a putative 407-nucleotide-long intron and give rise to two HEED protein isoforms, 535 and 494 amino acids in length. EED interacts in a highly specific manner, both in vitro and in vivo, with histone deacetylase (HDAC) proteins.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: EED
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat