To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:ADP-Ribosylation Factor-Like 4
- Antigen Symbol:ARL4
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Form:liquid
- Gene ID:10124
- Antigen Synonyms:ADP-ribosylation factor-like protein 4A|ADP ribosylation factor like 4A|ARL4A_HUMAN.|ARL4A|ARL 4
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10283-714
- Supplier no.:BS-12523R-CY7
ADP-ribosylation factors (ARFs) are highly conserved guanine nucleotide-binding proteins that enhance the ADP-ribosyltransferase activity of cholera toxin. ARF抯 are important in eukaryotic vesicular trafficking pathways and activating phospholipase D. ARL4 (ADP-ribosylation factor-like protein 4A) is a member of the ARF-like protein (ARL) subfamily of small GTPases. It contains a C terminal nuclear localization signal (NLS) region that interacts with Importin-? ARL4 localizes to the nucleus and is found in a variety of tissues, but is predominantly expressed in spermatogonia and Sertoli cells. It is most closely related to ARL6 and ARL7. Unlike ARFs, ARL4 does not activate the cholera toxin ADP-ribosyltranferase. ARL4 may play a role in neurogenesis during embryonic development and somitogenesis in the early stages of adult spermatogenesis.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ARL4A
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat