To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:ADP-Ribosylation Factor GTPase Activating Protein 3
- Antigen Symbol:ARFGAP3
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Form:liquid
- Gene ID:26286
- Antigen Synonyms:E2-EPF|UBE2S|ARFGAP1
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10280-676
- Supplier no.:BS-12513R-FITC
Specifications
About this item
ARFGAP1 (ADP-ribosylation factor GTPase-activating protein 1), ARFGAP2 (ADP-ribosylation factor GTPase-activating protein 2) and ARFGAP3 (ADP-ribosylation factor GTPase-activating protein 3) are GTPase-activating proteins (GAP) that are associated with the Golgi apparatus and interact with ADP-ribosylation factor 1 (ARF). These proteins promote hydrolysis of ARF-bound GTP and are required for the dissociation of coat proteins from Golgi-derived membranes and vesicles. Dissociation of the coat proteins is required for the fusion of these vesicles with target compartments. The activity of ARFGAP1, ARFGAP2 and ARFGAP3 is stimulated by phosphoinosides and inhibited by phosphatidylcholine. The genes encoding ARFGAP1, ARFGAP2 and ARFGAP3 map to human chromosomes 20q13.33, 11p11.2 and 22q13.2, respectively.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ARFGAP3
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat