To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Sulfatase Modifying Factor 1
- Antigen Symbol:SUMF1
- Clonality:Polyclonal
- Conjugation:Cy3®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Form:liquid
- Gene ID:285362
- Antigen Synonyms:pFGE|AI463102|FGE|AI851573|AA543204
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10279-330
- Supplier no.:BS-12366R-CY3
Specifications
About this item
SUMF1 is a 374 amino acid alternatively spliced protein that localizes to the lumen of the endoplasmic reticulum and belongs to the sulfatase-modifying factor family. Expressed ubiquitously with highest expression in liver, kidney and pancreas, SUMF1 exists as either a monomer, a homodimer or a heterodimer (with SUMF2) and functions to oxidize sulfatase cysteine residues to an active FGIy residue, thereby playing an important role in sulfatase activity. Defects in the gene encoding SUMF1 are the cause of multiple sulfatase deficiency (MSD), a heterogeneous disorder characterized by metachromatic leukodystrophy, mucopolysaccharidosis, chondrodysplasia punctata, hydrocephalus, ichthyosis, neurologic deterioration and developmental delay.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SUMF1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat