To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Craniofacial Development Protein 1
- Antigen Symbol:CFDP1
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Form:liquid
- Gene ID:10428
- Antigen Synonyms:SWC5|CENP-29|BUCENTAUR|BCNT|CP27|Yeti|p97
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10283-366
- Supplier no.:BS-12357R-FITC
Specifications
About this item
CFDP1 is a 299 amino acid protein that is involved in embryogenesis and normal cell function. When treated with CFDP1 peptide, mouse molar teeth increase in size, whereas treating cells with against CFDP1 shows an increase in the number of apoptotic cells and gradual tooth disintegration. CFDP1 is highly expressed in developing mouse teeth and is expressed at lower levels in liver, lung and heart. The gene encoding CFDP1 maps to human chromsome 16, in a region that has been associated with inherited craniofacial diseases, such as fanconi anemia type A. There are two isoforms of CFDP1 that are produced as a result of alternative splicing events.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CFDP1
Clonality: Polyclonal
Clone: 0
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: